Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BAZ2A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-V86
75932-924EA 518 USD InStock
75932-924
Anti-BAZ2A Rabbit Polyclonal Antibody
Antibodies
TIP 5 is the large subunit of the nucleolar remodeling complex NoRC. NoRC causes the repression of ribosomal gene transcription. It was demonstrated that histone deacetylation is involved in this repression and that TIP5 is associated with the histone deacetylase HDAC1 and mediates the deacetylation of histones in the vicinity of the rDNA promoter. The interaction of TIP5 and HDAC1, which is necessary for transcriptional repression, is established by the C-terminal PHD finger and bromodomain. Anti-TIP5 Antibody is ideal for research in Gene Expression.

Anti-TIP5 Antibody is suitable for Chromatin Immunoprecipitation and Western Blots. Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 250 kDa in the appropriate cell lysate or extract.

Type: Primary
Antigen: BAZ2A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty