Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TLR6 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FG2
75932-928EA 518 USD InStock
75932-928
Anti-TLR6 Rabbit Polyclonal Antibody
Antibodies
Toll-like receptors (TLRs) are evolutionarily conserved pattern-recognition molecules resembling the toll proteins that mediate antimicrobial responses in Drosophila. These proteins recognize different microbial products during infection and serve as an important link between the innate and adaptive immune responses. The TLRs act through adaptor molecules such as MyD88 and TIRAP to activate various kinases and transcription factors so the organism can respond to potential infection. TLR6 was first identified as a close homolog of TLR1, sharing 69% sequence identify. Like TLR1, TLR6 can form heterodimers with TLR2, and these TLR6:TLR2 dimers coordinate macrophage activation by Gram-positive bacteria and the yeast cell wall particle zymosan. Activation of these complexes not only initiates pro-inflammatory cascades, but also can lead to apoptotic responses.

Anti-TLR6 Antibody has been tested for use in ELISA, Western Blotting, Immunocytochemistry. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 92 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: TLR6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty