Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TMEM147 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FH5
75932-948EA 518 USD InStock
75932-948
Anti-TMEM147 Rabbit Polyclonal Antibody
Antibodies
TMEM147 is a recently identified protein that associates with Nicalin and NOMO, forming a complex that is thought to be similar to the gamma-secretase complex. TMEM147 localizes to the endoplasmic reticulum (ER), and in zebrafish, is expressed during early development. TMEM147 also interacts with the M3 muscarinic receptor (M3R) in the ER and is thought to negatively its expression and function.

Anti-TMEM147 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 25 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: TMEM147
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty