Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ANO1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FH6
75932-950EA 518 USD InStock
75932-950
Anti-ANO1 Rabbit Polyclonal Antibody
Antibodies
Calcium-activated chloride channels (CaCC) are present in many cell types and mediate physiological functions such as epithelial secretion, sensory signal transduction, and smooth muscle contraction. Subunits of these CaCC's include the transmembrane proteins TMEM16A and TMEM16B. TMEM16A is expressed in epithelial cells of the kidney and lung, pancreas, and sensory neurons and its mRNA is seen in foregut, airway epithelia, and tracheal smooth muscle; mice lacking TMEM16A fail to survive past ten days and show aerophagia and little weight gain. TMEM16A is also overexpressed or amplified in multiple cancers associated with poor survival such as oral cancers and gastrointestinal stromal tumors, suggesting the development of CaCC modulators may be a viable therapeutic strategy.

Anti-TMEM16A Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 114 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: ANO1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty