Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TMEM184C Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FJ1
75932-960EA 518 USD InStock
75932-960
Anti-TMEM184C Rabbit Polyclonal Antibody
Antibodies
Anaplastic thyroid cancer is one of the most lethal forms of cancer, but the precise carcinogenic mechanism has not been identified. TMEM184C, also known as TMEM34, was identified in a cDNA microarray analysis as being down-regulated in anaplastic thyroid cancers compared to normal thyroid tissues. TMEM184C protein expression was also lower in cell lines derived from these types of cancers compared to that of normal thyroid tissues or cell lines based on other types of thyroid cancers. Furthermore, transfection of TMEM34 into KTA2 cells led to the inhibition of cell growth, suggesting that TMEM184C might act as a tumor suppressor in anaplastic thyroid cancers.

Anti-TMEM184C Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 50 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: TMEM184C
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty