Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Transferrin Fluorescein Conjugated Rabbit Polyclonal Antibody (FITC)

Supplier: Rockland Immunochemicals
209-4234-0100
75933-050EA 488 USD InStock
75933-050
Anti-Transferrin Fluorescein Conjugated Rabbit Polyclonal Antibody (FITC)
Antibodies
Human transferrin is encoded by the TF gene and is an iron-binding blood plasma glycoprotein that controls the level of free iron in biological fluids. Human transferrin binds iron very tightly but reversibly. Human transferrin is the most important iron pool in mammals. Human transferrin has a molecular weight of around 80 kDa and contains 2 specific high-affinity Fe(III) binding sites. The affinity of Human transferrin for Fe(III) is extremely high but decreases progressively with decreasing pH below neutrality. Human Transferrin also plays a role in the immune system, creating environments low in iron for which many pathogenic bacteria are unable to thrive.

Anti-Human transferrin Fluorescein is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms.

Type: Primary
Antigen: Transferrin Fluorescein Conjugated
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty