Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TRIM28 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FP2
75933-110EA 518 USD InStock
75933-110
Anti-TRIM28 Rabbit Polyclonal Antibody
Antibodies
TRIM28, also known as KAP-1, is a member of the Transcriptional Intermediary Factor 1 (TIF1) subfamily and contains a RING finger, B box, Coiled coil, PHD/TTC, and bromodomain. TRIM28 is a corepressor for Kruppel-associated box (KRAB)-domain-containing zinc finger proteins and plays a critical role in early embryogenesis. TRIM28 acts as a transcriptional mediator by binding liganded nuclear receptors, including retinoic acid (RAR), retinoid X (RXR) and estrogen (ER) receptors. TRIM28 associates with both heterochromatin and euchromatin, causing gene silencing by both HP1 binding and histone deacetylation.

Anti-TRIM28 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 89 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: TRIM28
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty