Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TRIM33 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FP5
75933-120EA 518 USD InStock
75933-120
Anti-TRIM33 Rabbit Polyclonal Antibody
Antibodies
TRIM24 (TIF1alpha), TRIM28 (TIF1beta), and TRIM33 (TIF1gamma) are three related cofactors belonging to the tripartite motif superfamily that interact with distinct transcription factors (1-2). The structure of TRIM33 is similar to TRIM24 and TRIM28, exhibiting multiple domains (RING finger, coiled coil, B boxes, PHD/TTC, and bromodomain) (3). TRIM33 functions in cell differentiation and development, playing a role in differentiation of hematopoietic cells (3). It interacts with the Smad2/3 transcription factor in hematopoietic, mesenchymal, and epithelial cell types to mediate different transcriptional effects in response to TGF-beta (4). It acts as an E3 ubiquitin-protein ligase and promotes SMAD4 ubiquitination (5).

Anti-TRIM33 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 123 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: TRIM33
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty