Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNNI3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-X34
75933-148EA 530 USD InStock
75933-148
Anti-TNNI3 Rabbit Polyclonal Antibody
Antibodies
Troponin I (TnI) is 1 of 3 subunits, along with troponin C (TnC) and Troponin T (TnT) of troponin complex found in cardiac (cTnI) and fast skeletal (fsTnI) muscle. Protein kinase C-mediated phosphorylation of cardiac myofilament proteins has been shown to depress the actomyosin interaction and may be important during heart failure. Studies indicate that phosphorylation of Ser43 and Ser45 of cardiac troponin I plays a substantial role in the PKC-mediated depression. Troponin I pS43 Antibody is ideal for researchers interested in cancer research.

Anti-Phospho-Ser43 Troponin (cardiac) Antibody is stored with 100 μg per ml BSA. It is suitable for Western Blots and is specific for the ~25 kDa cardiac troponin I protein phosphorylated at Ser 43. Immunolabeling is greatly decreased with lambda-phosphatase treatment. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: TNNI3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty