Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TSC22D3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FR2
75933-182EA 518 USD InStock
75933-182
Anti-TSC22D3 Rabbit Polyclonal Antibody
Antibodies
The TSC22 domain family member 3 protein (TSC22D3) is a leucine zipper protein that functions as a transcriptional regulator. The expression of TSC22D3 is stimulated by glucocorticoids and IL-10 and is thought to play a key role in the anti-inflammatory and immunosuppressive effects of these molecules. TSC22D3 can physically interact with and inhibit the activities of key inflammatory signaling mediators such NF-kB and AP-1. TSC22D3 functions as a transcriptional co-activator for various nuclear receptors and NF-kB. It has also been shown to be involved in the differentiation of mesenchymal stem cells towards osteoblasts and bone formation.

Anti-TSC22D3 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 15 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: TSC22D3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty