Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TSLP Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
209-401-H48
75933-190EA 518 USD InStock
75933-190
Anti-TSLP Rabbit Polyclonal Antibody
Antibodies
Thymic Stromal Lymphopoietin (TSLP) is a hematopoietic cytokine. TSLP is mainly expressed in the heart, liver and prostate. TSLP sends signals through a heterodimeric receptor complex, containing the TSLP-R and the IL-7Rα chain to induce the release of T cell-attracting chemokines from monocytes. It can induce allergic inflammation by directly activating mast cells. It also promotes T helper type 2 (TH2) cell responses associated with immunity, including atopic dermatitis and asthma, and can promote TH2 cytokine-associated inflammation by directly promoting the effector functions of CD4+ TH2 cells, basophils and other granulocyte populations while simultaneously limiting the expression of dendritic cells derived proinflammatory cytokines and promoting regulatory T cell responses in peripheral tissues. Recombinant human TSLP is a non-glycosylated protein with a molecular weight of 15 kDa. Anti-TSLP antibody is ideal for researchers interested in cytokines and stem cell research.

TSLP purified antibody has been tested for use in ELISA and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 18 kDa in size corresponding to the human TSLP protein by western blotting in appropriate cell lysate or extract.

Type: Primary
Antigen: TSLP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty