Would you like to log in again?
Turkey Serum (Lyophilized), Rockland Immunochemicals
Supplier: Rockland Immunochemicals
D313-05
75933-280EA 177 USD InStock
75933-280
Turkey Serum (Lyophilized), Rockland Immunochemicals
Sera (Serum)
Lyophilized Non-Sterile Serum
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques
Suitable for use as a control or diluent in assays and most other immunological methods.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty