Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TXNIP Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FT0
75933-288EA 518 USD InStock
75933-288
Anti-TXNIP Rabbit Polyclonal Antibody
Antibodies
Thioredoxin-interacting protein (TXNIP) belongs to the arrestin family and plays a critical role in the antioxidant defense mechanisms of hematopoietic cells by activating the p53 pathway during oxidative stress (1,2). It functions as a transcriptional repressor and acts as an oxidative stress mediator by inhibiting thioredoxin activity (2). TXNIP expression is reduced in many types of tumors, and TXNIP overexpression inhibits tumor growth by blocking cell-cycle progression (3). It has recently reported that TXNIP deficiency correlates with a high incidence of hepatocellular carcinoma (HCC) (4). TXNIP and p53 interactions could potentially be a therapeutic target for oxidative stress-related diseases such as hematopoietic malignancies and metabolic diseases (5).

Anti-TXNIP Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 44 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: TXNIP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty