Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-UBIAD1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FT7
75933-392EA 518 USD InStock
75933-392
Anti-UBIAD1 Rabbit Polyclonal Antibody
Antibodies
The UbiA prenyltransferase domain containing 1 (UBIAD1) protein , also known as TERE1, was initially identified as a down-regulated gene in transitional cell carcinoma of the bladder (1). Recently it has been shown to bind the cholesterol carrier APOE and modulate cellular cholesterol levels (2). Mutations in the UBIAD1 gene can cause Schnyder crystalline corneal dystrophy, an autosomal dominant disease characterized by progressive opacification of the cornea resulting from the local accumulation of lipids (3).

Anti-UBIAD1 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 37 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: UBIAD1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty