Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-UHRF1BP1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FU1
75933-426EA 518 USD InStock
75933-426
Anti-UHRF1BP1 Rabbit Polyclonal Antibody
Antibodies
The Ubiquitin-like containing PHD and RING finger domains 1-binding protein 1 (UHRF1BP1), also known as ICBP90, is a transcription and cell cycle regulator. It specifically binds to the histone H3 N-terminal tail when methylated on K9 by two functional domains, a PHD finger that defines the binding specificity and an SRA (SET- and RING-associated) domain that promotes binding activity. UHRF1BP1 is required for proper heterochromatin formation in mammalian cells. Furthermore, UHRF1BP1 is thought to be a pivotal target for the ERK1/2 signaling pathway to control the proliferation of Jurkat T cells.

Anti-UHRF1BP1 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 160 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: UHRF1BP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty