Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ULK1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FU4
75933-430EA 518 USD InStock
75933-430
Anti-ULK1 Rabbit Polyclonal Antibody
Antibodies
ULK1, also known as ATG1, is a key serine/threonine protein kinase probably acting at the most upstream step of autophagosome formation. Knockout of ULK1 results in a severe defect in the autophagy pathway. ULK1 is highly conserved among eukaryotes, and are the Unc-51-like kinases, ULK1 and ULK2 in mammals. ULK1 is ubiquitously expressed and involved in autophagy in response to starvation. It is the target of the TOR kinase signaling pathway that regulates autophagy through the control of phosphorylation status of ATG13. ULK1 also plays a role early in neuronal differentiation.

Anti-ULK1 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 113 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: ULK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty