Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ULK3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FU6
75933-434EA 518 USD InStock
75933-434
Anti-ULK3 Rabbit Polyclonal Antibody
Antibodies
ULK3 belongs to the Ser/Thr protein kinase superfamily and plays a role in the ATP-dependent phosphorylation of target proteins (1). Knockout of ULK genes results in a severe defect in the autophagy pathway (2). ULK3, like the other Unc-51-like kinases such as ULK1, ULK2 and ULK4, is highly conserved among eukaryotes (3). ULK3 has been shown to be a positive regulator of the Hedgehog signaling pathway by enhancing GLI1 and GLI2 transcriptional activity (4). Furthermore, ULK3 can also interact with SUFU, a protein required for the negative regulation of GLI proteins; this interaction blocks the autophosphorylation of ULK3 and blocks its ability to regulate the GLI proteins (5).

Anti-ULK3 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 53 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: ULK3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty