Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ULK4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FU7
75933-436EA 518 USD InStock
75933-436
Anti-ULK4 Rabbit Polyclonal Antibody
Antibodies
ULK4 belongs to the Ser/Thr protein kinase superfamily and plays a role in the ATP-dependent phosphorylation of target proteins (1). Knockout of ULK genes results in a severe defect in the autophagy pathway (2). ULK4, like the other Unc-51-like kinases such as ULK1, ULK2 and ULK3, is highly conserved among eukaryotes (3). ULK4 has been implicated as a gene involved in the development of hydrocephalus (3). This neurologic disorder is being used in animal models to elucidate factors responsible for the excessive accumulation of cerebrospinal fluid in hydrocephalic humans (4).

Anti-ULK4 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry, and Immunofluorescence. Expect a band at approximately 142 kDa in Western Blots using positive control NIH3T3 whole cell lysates, human brain tissue for IHC, or other specific cell lysates and tissues. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: ULK4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty