Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-UNC93A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FU8
75933-442EA 518 USD InStock
75933-442
Anti-UNC93A Rabbit Polyclonal Antibody
Antibodies
Unc93a, a human homolog of the C. elegans Unc93 gene, was initially identified by the sequencing of chromosome 6q27 at D6S193 as loss at this region correlated with sporadic ovarian cancer. Further analysis however, showed no tumor suppressor role for Unc93a. Unlike Unc93b, another homolog of the C. elegans Unc93 gene that is crucial for Toll-like receptor (TLR) signaling, the function of Unc93a is unknown. It is expressed in testis, small intestine, spleen, prostate and ovary.

Anti-Unc93a Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 50 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: UNC93A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty