Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ANKRD27 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FV9
75933-464EA 518 USD InStock
75933-464
Anti-ANKRD27 Rabbit Polyclonal Antibody
Antibodies
The VPS9 ankyrin repeat protein (VARP) binds to the Rab21, a guanine nucleotide exchange factor that plays an essential role in endocytic trafficking. VARP localizes to early endosomes and is thought to regulate endosome dynamics. VARP also interacts with TI-VAMP/VAMP7, a vesicular SNARE that mediates an exocytic pathway that is crucial to neurite growth. Depletion of VARP by RNA interference impairs neurite growth, suggesting that VARP is a positive regulator of neurite growth.

Anti-VARP Antibody has been tested for use in ELISA, and Immunohistochemistry. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 117 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: ANKRD27
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty