Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VASH1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FW1
75933-508EA 518 USD InStock
75933-508
Anti-VASH1 Rabbit Polyclonal Antibody
Antibodies
VASH1 was originally identified as an endothelium-derived vascular endothelial growth factor (VEGF)-inducible angiogenesis inhibitor that acts in a negative feedback manner. VASH1 mRNA is expressed brain, heart, kidney and placenta in the adult, with robust expression in various organs during embryonic development. VASH1 expression in tumor cells and tumor vasculature is silenced by methylation by increased expression of Zeste homolog 2 (EZH2). The increase of EZH2 expression is induced by VEGF stimulation, leading to poor clinical prognosis. Recombinant adenovirus expressing VASH prevented tumor angiogenesis and inhibited tumor growth, suggesting that it may be a potentially valuable antitumor therapy in the clinic.

Anti-VASH1 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 41 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: VASH1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty