Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VDAC1 Mouse Monoclonal Antibody (RPE) [Clone: S152B-23]

Supplier: Rockland Immunochemicals
200-308-W70
75933-544EA 539 USD InStock
75933-544
Anti-VDAC1 Mouse Monoclonal Antibody (RPE) [Clone: S152B-23]
Antibodies
Voltage-dependent anion-selective channel protein 1 (also known as VDAC, VDAC1 or outer mitochondrial membrane protein porin 1) is the the outer mitochondrial membrane receptor for hexokinase and BCL2L1. VDAC forms a channel through the mitochondrial membrane and is involved in small molecule diffusion, cell volume regulation and apoptosis. VDAC may participate in the formation of the permeability transition pore complex (PTPC), which is responsible for the release of mitochondrial products that triggers apoptosis. Anti-VDAC1 is ideal for research in Cell Biology and Apoptosis.

Anti-VDAC1 R-PE Conjugated Antibody is suitable for Western Blots, Immunohistochemistry and Immunocytochemistry. Expect a band approximately ~30kDa on specific lysates or tissues. Does not cross react with VDAC2 or VDAC3 (based on KO validation results). Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: VDAC1
Clonality: Monoclonal
Clone: S152B-23
Conjugation: RPE
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty