Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KDR Mouse Monoclonal Antibody [Clone: FLK-6]

Supplier: Rockland Immunochemicals
200-301-P49
75933-548EA 528 USD InStock
75933-548
Anti-KDR Mouse Monoclonal Antibody [Clone: FLK-6]
Antibodies
VEGF, a homodimeric glycoprotein of relative molecular mass 45,000, is the only mitogen that specifically acts on endothelial cells. The importance of VEGF and its receptor system in tumor growth and intervention in this system may provide promising approaches to cancer therapy. VEGF receptor 2 is a member of a receptor tyrosine kinase family.Like other growth factor receptors, upon ligand binding VEGF receptor 2 dimerises and is autophosphorylated on multiple tyrosine residues. This antibody is suitable for researchers interested in cell signaling and cancer research.

Anti-VEGFR2 is useful for Immunohistochemistry -F. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~151.5 kDa corresponding to the appropriate cell lysate or extract.

Type: Primary
Antigen: KDR
Clonality: Monoclonal
Clone: FLK-6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty