Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VEZT Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FW3
75933-552EA 518 USD InStock
75933-552
Anti-VEZT Rabbit Polyclonal Antibody
Antibodies
Vezatin (VEZT) is a transmembrane protein which localizes to adherens junctions and binds to myosin VIIA and the cadherin-catenins complex (1). Expression of this gene in gastric cancer tissues is decreased due to hypermethylation of the promoter. Expression of this gene may also be inhibited by binding of a specific microRNA to a target sequence in the 3' UTR of the transcripts (2). VEZT has also been shown to be required for morphogenesis of the preimplantation mouse embryo (3).

Anti-VEZT Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 89 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: VEZT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty