Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VIM Mouse Monoclonal Antibody [Clone: 2D1]

Supplier: Rockland Immunochemicals
200-301-E54
75933-556EA 528 USD InStock
75933-556
Anti-VIM Mouse Monoclonal Antibody [Clone: 2D1]
Antibodies
Vimentin is the major protein subunit of the 10nm or intermediate filaments (IFs) found in many kinds of mesenchymal and epithelial cells as well as developing neuronal and astrocytic precursor cells in the CNS. Vimentin is thought to be critically involved in lymphocyte adhesion and transmigration. Copolymers are frequently formed between vimentin and other IFs, such as GFAP (in many kinds of astrocytes), desmin (in muscle cells) and neurofilament proteins (in developing neurons). Antibodies to vimentin are useful in studies of stem cells and generally to reveal the filamentous cytoskeleton. Recent studies suggest that vimentin affects prostate cancer cells motility and invasiveness. Vimentin Antibody is ideal for researchers interested in neuroscience research.

Anti-Vimentin Antibody is suitable for Western Blots and Immunofluorescence, and it is specific for the ~50 kDa vimentin protein. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: 2D1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty