Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VSNL1 Mouse Monoclonal Antibody [Clone: 2D11]

Supplier: Rockland Immunochemicals
200-301-X36
75933-580EA 528 USD InStock
75933-580
Anti-VSNL1 Mouse Monoclonal Antibody [Clone: 2D11]
Antibodies
Visinin-like protein 1 (VSNL1), also known as VILIP1, is a calcium sensor protein expressed exclusively in neurons. Highest levels of VSNL1 expression are found in cerebellar Purkinje cells. VSNL1 has been implicated in the modulation of cell signaling cascades via regulation of adenyl cyclase activity. Additionally, VSNL1 has been associated with amyloid plaques and neurofibrillar tangles in Alzheimer’s disease. Visinin-Like Protein 1 Antibody is ideal for researchers interested in neuroscience research.

Anti-Visinin-Like Protein 1 Antibody is suitable for Western Blots and Immunofluorescence, and it is specific for the ~22 kDa VSNL protein. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: VSNL1
Clonality: Monoclonal
Clone: 2D11
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty