Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VNN3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FX1
75933-590EA 518 USD InStock
75933-590
Anti-VNN3 Rabbit Polyclonal Antibody
Antibodies
The vanin family is a novel group of ectoenzymes that function in tissue repair and plays a role in oxidative-stress response. As both secreted and membrane proteins, the vanin family members have been implicated as therapeutic targets in inflammatory disease. VNN3 (vascular non-inflammatory molecule 3), also known as Vanin3, is a 501 amino acid GPI-anchored amidohydrolase that is widely expressed and is found at highest levels in blood and liver. Induced by Th17 / Th1 type cytokines, VNN3 converts pantetheine into pantothenic acid. Containing one CN hydrolase domain, VNN3 is encoded by a gene that maps to human chromosome 6q23.2.

Anti-VNN3 Antibody has been tested for use in ELISA, Western Blotting, and Immunohistochemistry. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 56 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: VNN3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty