Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-XKR8 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GA1
75933-656EA 518 USD InStock
75933-656
Anti-XKR8 Rabbit Polyclonal Antibody
Antibodies
The XK Kell blood group complex subunit-related family member 8 (XKR8) is a member of a family of proteins that promote the phagocytosis of dying cells by altering the phospholipid distribution of the plasma membrane (1). XKR8 is thought to be a “scramblase”, an enzyme that eliminates the asymmetry of phospholipid distribution in the plasma membrane. In apoptotic cells, XKR8 promotes the cell-surface exposure of phosphatidylserine (PtdSer) which serves as a signal for phagocytosis (1,2).

Anti-XKR8 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 45 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: XKR8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty