Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ZNF219 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GD3
75933-716EA 518 USD InStock
75933-716
Anti-ZNF219 Rabbit Polyclonal Antibody
Antibodies
ZFP219 is a developmentally regulated member of the Kruppel-like zinc finger gene family that is thought to function as a transcriptional repressor. Yeast two-hybrid screening showed association with Sox9, a transcription factor that is essential for chondrogenesis. ZFP219 is specifically expressed in the developing limb buds and colocalizes with Sox9 in the nucleus. Knockdown of ZFP219 expression decreased Sox9-induced mRNA expression, and a dominant-negative mutant of ZFP219 inhibited Bmp2-induced chondrocyte differentiation, suggesting that ZFP219 plays an important role as a transcriptional partner of Sox9 in the regulation of chondrocyte differentiation.

Anti-ZFP219 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 77 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: ZNF219
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty