Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Zic1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-159S
75933-726EA 148 USD InStock
75933-726
Anti-Zic1 Rabbit Polyclonal Antibody
Antibodies
Anti Zic-1 Antibody recognizes the product of the Zic-1 gene, that encodes a zinc finger protein which is expressed in the developing or matured central nervous system in a highly restricted manner. Zic-1 is expressed in granule cells that make synaptic contact with Purkinje cells. Clearly Zic-1 is a gene critical to cerebellar pattern formation. The expression of Zic genes is first detected at gastrulation and at neurulation, becomes restricted to the dorsal neural ectoderm and the dorsal paraxial mesoderm. Zic-2 and Zic-3 are highly similar genes, especially in their product's zinc finger motif and by comparison of their genomic organization in that they share common exon-intron boundaries and belong to the same gene family. By comparison in function, Zic-2 is essential for the formation of the brain and Zic-3 is important for right and left axis formation. The Zic-1 gene has been mapped to chromosome 9 in mouse. The 5' flanking region of the Zic-1 gene contains a region-specific enhancer determined to be essential in in vivo and in vitro deletion analysis. The temporal profile of mRNA expression differs for each of the Zic gene products. The Drosophila odd-paired gene is highly homologous to the Zic gene family.

Type: Primary
Antigen: Zic1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty