Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ZMIZ1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GD8
75933-728EA 518 USD InStock
75933-728
Anti-ZMIZ1 Rabbit Polyclonal Antibody
Antibodies
ZIMP10, also known as ZMIZ1, is a novel PIAS (protein inhibitor of activated signal transducer and activator of transcription)-like protein initially identified as a transcriptional co-activator of the androgen receptor (AR). ZIMP10 and the related protein ZIMP7 interact with PIAS3 and enhances AR-mediated transcription. Later experiments showed that ZIMP10 is also a co-activator of the p53 tumor suppressor. Mice deficient in ZIMP10 result in embryonic lethality by E10.5; these embryos reveal severe defects in the reorganization of the yolk sac vascular plexus, indicating that ZIMP10 plays an important role in proper vascular development.

Anti-ZIMP10 Antibody has been tested for use in ELISA, Western Blotting, Immunocytochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 115 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: ZMIZ1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty