Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Slc39a13 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GE5
75933-742EA 518 USD InStock
75933-742
Anti-Slc39a13 Rabbit Polyclonal Antibody
Antibodies
The zinc transporter ZIP13, also known as SLC39A13, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. The zinc transporter family is divided into four subfamilies (I, II, LIV-1 and gufA). ZIP13 is a multipass membrane protein that belongs to the ZIP transporter subfamily LIV-1. Mutations in ZIP13 have recently been shown to cause a spondylocheiro dysplastic form of Ehlers-Danlos syndrome (SCD-EDS), a generalized skeletal dysplasia involving mainly the spine with clinical abnormalities of the hands in addition to EDS-like features. Other experiments have shown that ZIP13 is required for proper connective tissue development and is involved in BMP/TGF- signaling pathways.

Anti-ZIP13 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 39 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: Slc39a13
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty