Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SLC39A6 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GF1
75933-752EA 530 USD InStock
75933-752
Anti-SLC39A6 Rabbit Polyclonal Antibody
Antibodies
The zinc transporter ZIP6, also known as SLC39A6, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. ZIP6 was initially identified as LIV-1, an estrogen-regulated gene that has been implicated in metastatic breast cancer. Elevated ZIP6 expression has also been reported in human cervical cancer and the HeLa cell line; down-regulation of ZIP6 expression in HeLa by RNAi inhibited cell proliferation, colony formation, migration and invasiveness, as well as decreasing Snail and Slug levels, suggesting ZIP6 plays a regulatory role on the ERK1/2-Snail/Slug pathway.

Anti-ZIP6 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 85 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: SLC39A6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty