Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ZNF346 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-GG1
75933-766EA 518 USD InStock
75933-766
Anti-ZNF346 Rabbit Polyclonal Antibody
Antibodies
ZNF346, also known as JAZ (just another zinc-finger protein), is a nucleolar, zinc finger protein which preferentially binds to double-stranded (ds) RNA or RNA/DNA hybrids with high affinity via C2H2 zinc fingers. ZNF346 contains four CH-type zinc finger motifs that are connected by long (28-38) amino acid linker sequences. ZNF346 is expressed in all tissues tested and localizes to the nucleus, primarily the nucleolus. ZNF346 is exported by exportin-5 but translocates back into nuclei by a facilitated diffusion mechanism. ZNF346 interacts with ILF3 in an RNA-independent manner and may be involved in cell growth and survival.

Anti-ZNF346 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 33 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: ZNF346
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty