Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KDM1B Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-W43
75933-792EA 487 USD InStock
75933-792
Anti-KDM1B Rabbit Polyclonal Antibody
Antibodies
KDM1B is a histone demethylase that demethylates lysine 4 of histone H3, a specific tag for epigenetic transcriptional activation, thereby acting as a corepressor. KDM1B is able to demethylate both mono- and di-methylated lysine 4 of histone H3, but has no effect on tri-methylated lysine 4, mono-, di- or tri-methylated lysine 9, mono-, di- or tri-methylated lysine 27, mono-, di- or tri-methylated lysine 36 of histone H3, or on mono-, di- or tri-methylated lysine 20 of histone H4. KDM1B is also required for de novo DNA methylation of a subset of imprinted genes during oogenesis. It acts by oxidizing the substrate by FAD to generate the corresponding imine that is subsequently hydrolyzed. Anti-KDM1B is ideal for research in Epigenetics and Gene Expression.

Anti-KDM1B Antibody is suitable for ELISA. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: KDM1B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty