Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Ly6al Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-P88
75933-832EA 487 USD InStock
75933-832
Anti-Ly6al Rabbit Polyclonal Antibody
Antibodies
Ly6al, lymphocyte antigen 6 complex, locus A-like, is also known as  Ly6a, Ly6a_predicted. This antibody is suitable for researchers interested in stem cell research.

Anti-Lys6al is useful for Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~14.2 kDa corresponding to the appropriate cell lysate or extract.

Type: Primary
Antigen: Ly6al
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty