Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MALT1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-CQ0
75933-856EA 518 USD InStock
75933-856
Anti-MALT1 Rabbit Polyclonal Antibody
Antibodies
MALT1 was initially identified as a novel gene that was recurrently rearranged in t(11;18)(q21;q21) mucosa-associated lymphoid tissue lymphomas along with the apoptosis inhibitor protein c-IAP2 (1). MALT1, along with the proteins CARMA1 and Bcl10 form an NF-kappaB-activating complex, termed the CBM signalsome, that acts downstream of lymphocyte antigen receptors as well as many other non-lymphoid cell-surface receptors that play a role in multiple cellular functions (2,3). MALT1 has proteolytic activity, and this activity is critical for full NF-kappaB response in T cell activation (4).

Anti-MALT1 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 92 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: MALT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty