Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LAF Goat Polyclonal Antibody (BAC)

Supplier: Rockland Immunochemicals
200-106-230-0100
75933-862EA 498 USD InStock
75933-862
Anti-LAF Goat Polyclonal Antibody (BAC)
Antibodies
Anti-Maltose Phosphorylase recognizes the protein maltose phosphorylase which belongs to the glycosyltransferases family. Maltose phosphorylase catalyzes the reaction in which maltose and inorganic phosphate are converted to D-glucose and beta-D-glucose 1-phosphate.

Anti-Maltose Phosphorylase antibody has been assayed against 1.0 µg of Maltose Phosphorylase in a standard capture ELISA using Peroxidase Conjugated Streptavidin #S000-03 and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid]) code # ABTS-100 as a substrate for 30 minutes at room temperature.  A working dilution of 1:4,000 to 1:20,000 of the reconstitution concentration is suggested for this product.

Type: Primary
Antigen: LAF
Clonality: Polyclonal
Clone:
Conjugation: BAC
Epitope:
Host: Goat
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty