Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Mitofusin-2 Mouse Monoclonal Antibody (ALP) [Clone: S153-5]

Supplier: Rockland Immunochemicals
200-305-W71
75933-888EA 539 USD InStock
75933-888
Anti-Mitofusin-2 Mouse Monoclonal Antibody (ALP) [Clone: S153-5]
Antibodies
Mitofusin-2 (Mfn2) is a membrane protein of the mitochondria and is important in mitochondrial fusion in mammalian cells. Mfn2 activity has also been implicated in mitochondrial metabolism and the loss of Mfn2 may be the reason for certain metabolic modifications that are seen in individuals with obesity. Mfn2 has been associated with maintaining the membrane potential of the mitochondria. Mfn2 has been shown to hinder mitochondrial antiviral signaling by inhibiting production of type 1 interferons. Mfn2 is a p53-inducible target and involved in cell proliferation, apoptosis promotion and tumor suppression. Anti-Mitofusin-2 is ideal for research in Metabolic Processes and Hemostasis.

Anti-Mitofusin 2 Alkaline Phosphatase Conjugated Antibody is suitable for Western Blots, Immunohistochemistry and Immunocytochemistry. Expect a band approximately ~90kDa on specific lysates or tissues. Does not cross react with Mitofusin 1. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: Mitofusin-2
Clonality: Monoclonal
Clone: S153-5
Conjugation: ALP
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty