Would you like to log in again?
Mouse IgG
Supplier: Rockland Immunochemicals
010-0102-0005
75933-924EA 262 USD InStock
75933-924
Mouse IgG
Proteins and Peptides
MOUSE IgG Whole Molecule
  • control or standard reagent
  • SDS, Western Blotting, and ELISA experiments
Mouse IgG purified protein (Immunoglobulin G) are antibody molecules. Mouse IgG is composed of four peptide chains — two heavy chains and two light chains. Mouse IgG has two antigen binding sites. Other Immunoglobulins may be described in terms of polymers with the IgG structure considered the monomer. Mouse IgG typically constitutes 75% of serum immunoglobulins. Mouse IgG molecules are synthesized and secreted by plasma B cells. Ideal as a negative control for Flow Cytometry, Western blotting, immunoprecipitation and immunohistochemistry applications.
Mouse IgG whole molecule can be utilized as a control or standard reagent in SDS, Western Blotting, and ELISA experiments.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty