Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Neuroligin 4 Mouse Monoclonal Antibody (PerCP) [Clone: S98-7]

Supplier: Rockland Immunochemicals
200-365-W81
75933-978EA 539 USD InStock
75933-978
Anti-Neuroligin 4 Mouse Monoclonal Antibody (PerCP) [Clone: S98-7]
Antibodies
This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses. The encoded protein interacts with discs, large (Drosophila) homolog 4 (DLG4). Mutations in this gene have been associated with autism and Asperger syndrome. Two transcript variants encoding the same protein have been identified for this gene. Anti-Neuroligin-4 is ideal for research in Neuroscience, including synaptic transmission and GABAergic inhibition, as well as Cell Adhesion.

Anti-Neuroligin 4 PerCP Conjugated Antibody is suitable for Western Blots, Immunofluorescence (Primary) and Immunocytochemistry. Expect a band approximately ~125-130kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: Neuroligin 4
Clonality: Monoclonal
Clone: S98-7
Conjugation: PerCP
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty