Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NUMB Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DF1
75934-018EA 518 USD InStock
75934-018
Anti-NUMB Rabbit Polyclonal Antibody
Antibodies
NUMB, the mammalian homolog to the Drosophila asymmetric cell fate determinant NUMB, is thought to share several features molecular mechanisms in mammalian cells, generating asymmetric cell divisions during neurogenesis in vertebrate development as well as in hematopoietic stem cells. NUMB has been shown to inhibit Notch signaling, and is itself regulated by ubiquitinylation by MDM2. NUMB has also been shown to help activate the tumor suppressor p53, suggesting that loss of NUMB in cancerous cells would not only activate the potential oncogene Notch, but diminish the tumor suppressing effect of p53.

Anti-NUMB Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 71 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: NUMB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty