Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PAPOLA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
100-401-V99
75934-086EA 487 USD InStock
75934-086
Anti-PAPOLA Rabbit Polyclonal Antibody
Antibodies
PAPOLA is an RNA polymerase that generates the poly(A) tail of RNA’s by adding AMP monomers to the 3’-end. PAPOLA may also be required for the endoribonucleolytic cleavage reaction at some polyadenylation sites. Anti-PAPOLA Antibody is ideal for research in Gene Expression, Transcription and Translation.

Anti-PAPOLA Antibody is suitable for ELISA and Western Blots. Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 83 kDa in the appropriate cell lysate or extract.

Type: Primary
Antigen: PAPOLA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty