Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PML Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-X44
75934-104EA 487 USD InStock
75934-104
Anti-PML Rabbit Polyclonal Antibody
Antibodies
PML is a transcription factor and tumor supressor which belongs to the tripartite motif (TRIM) family. This motif contains three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. PML regulates the response of p53 to oncogenic signals. It is often involved in a chromosomal translocation between chromosomes 15 and 17 which results in a PML/RARalpha fusion protein and is associated with most cases of acute promyelocytic leukemia. Anti-PML Antibody is ideal for research in Gene Expression, Cell Cycle Control and Cancer.

Anti-PML Antibody is suitable for ELISA and Western Blots. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 115 and 60 kDa in the appropriate cell lysate or extract.

Type: Primary
Antigen: PML
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty