Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-POLR2A Mouse Monoclonal Antibody

Supplier: Rockland Immunochemicals
200-301-U99
75934-110EA 528 USD InStock
75934-110
Anti-POLR2A Mouse Monoclonal Antibody
Antibodies
RNA polymerase II (pol II) is a key enzyme in the regulation and control of gene transcription. It is able to unwind the DNA double helix, synthesize RNA, and proofread the result. Pol II is a complex enzyme, consisting of 12 subunits, of which the B1 subunit is the largest. Together with the second largest subunit, B1 forms the catalytic core of the RNA polymerase II transcription machinery. Anti-Pol II Antibody is ideal for research in Gene Expression, Transcription and Genetics.

Anti-Pol II Antibody is suitable for Chromatin Immunoprecipitation Sequencing, Chromatin Immunoprecipitation, ELISA, Immunofluorescence and Western Blots. Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 225 kDa in the appropriate cell lysate or extract.

Type: Primary
Antigen: POLR2A
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty