Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TTR Chicken Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-901-FM9
75934-280EA 518 USD InStock
75934-280
Anti-TTR Chicken Polyclonal Antibody
Antibodies
Transthyretin is a tetrameric carrier protein that transports thyroid hormones in the plasma and cerebrospinal fluid, and retinol (vitamin A) in the plasma. More than 80 different mutations in this gene have been reported; most mutations are related to amyloid deposition, affecting predominantly peripheral nerve and/or the heart. The diseases caused by mutations include familial amyloidotic polyneuropathy, euthyroid hyperthyroxinemia, amyloidotic vitreous opacities, cardiomyopathy, oculoleptomeningeal amyloidosis, meningocerebrovascular amyloidosis, and carpal tunnel syndrome. It has also been suggested that Transthyretin plays an important role in the maintenance of normal cognitive processes during aging, neuropeptide processing and nerve regeneration. It has also been linked to several pathological conditions including Parkinson's disease, schizophrenia, and depression.

Anti-Transthyretin Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 16 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: TTR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty