Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ACP5 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FN0
75934-282EA 518 USD InStock
75934-282
Anti-ACP5 Rabbit Polyclonal Antibody
Antibodies
TRAP, also known as uteroferrin, is an iron containing, glycosylated, acid phosphatase. It is the most basic of the acid phosphatases and is the only form not inhibited by L(+)-tartrate. Along with the related protein ACP2, TRAP mediates the removal of mannose 6-phosphate residues from proteins targeted to lysosomes. TRAP is present in brain at low levels, but is expressed at a much higher level in liver.

Anti-TRAP Antibody has been tested for use in ELISA, Western Blotting and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 37 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: ACP5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty