Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TRB3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-FN1
75934-284EA 518 USD InStock
75934-284
Anti-TRB3 Rabbit Polyclonal Antibody
Antibodies
TRB3 was initially identified as a novel kinase-like gene that was overexpressed in multiple human tumors and regulated by hypoxia (1). In the liver, TRB3 binds and inhibits AKT activity, leading to impaired insulin signaling (2), whereas in adipose tissue TRB3 is involved in the regulation of fatty acid oxidation through ubiquitination of Acetyl-CoA carboxylase (3). Endoplasmic reticulum (ER) stress induces TRB3 expression in skeletal muscle, leading to a decrease in insulin signaling and glucose uptake, suggesting that inhibition of TRB3 expression may be a potential therapeutic treatment for managing insulin resistance (4).

Anti-TRB3 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 40 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: TRB3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty