Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HEPACAM2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BQ9
75934-384EA 518 USD InStock
75934-384
Anti-HEPACAM2 Rabbit Polyclonal Antibody
Antibodies
HEPACAM2 (Hepatocyte cell adhesion molecule 2), a type I N-linked transmembrane glycoprotein, belongs to the immunoglobulin superfamily. The exact function of HEPACAM2 is currently unknown, but the related protein HEPACAM forms cishomodimers on the cell surface to regulate the cell adhesion and may inhibit cell growth through suppression of cell proliferation. HEPACAM and HEPACAM2 mRNA are differentially regulated in canine tumors, with HEPACAM2 mRNA showing increased levels in adenomas, but decreased levels in metastatic carcinomas compared to normal tissues, while HEPACAM protein levels decreased in adenomas. It is therefore likely that HEPACAM2 plays a different role than HEPACAM in the development and progression to tumors.

Anti-HEPACAM2 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 51 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: HEPACAM2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty