Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HES5 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BR0
75934-390EA 518 USD InStock
75934-390
Anti-HES5 Rabbit Polyclonal Antibody
Antibodies
HES5 is a member of the HES family of transcriptional repressors, which are the mammalian homologs of the Drosophila Hairy and Enhancer of Split and contain basic Helix-loop-helix (bHLH) and Orange domains (1). The HES5 gene is a downstream target of the Notch signaling pathway and its expression is downregulated during human cartilage differentiation (2). HES5 can directly repress transcription of the E3 ligase FBW7-beta, creating a feedback loop that modulates Notch-mediated intestinal and neural stem cell fate decisions (3).

Anti-HES5 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 18 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: HES5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty